Learn More
Biomatik Corporation Human Tissue Polypeptide Specific Antigen (TPS) CLIA Kit
Supplier:  Biomatik Corporation EKU08936
Encompass_Preferred
Human Tissue Polypeptide Specific Antigen (TPS) CLIA Kit for detecting TPS in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05783. Gene ID: 3875. Target Synonyms: Specific M3 Epitope Of TPA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8588