Learn More
ACROBiosystems Human TL1A-DR3 Inhibition Kit (TR-FRET), 100 tests
Catalog No. NC3731147
Encompass_Preferred
Human TL1A-DR3 Inhibition Kit (TR-FRET) is a homogeneous, no-wash competition assay for rapid screening of inhibitors that block TL1A binding to DR3. The assay uses time-resolved fluorescence resonance energy transfer for sensitive, high-throughput detection and produces results within 0.5-1 hour.
  • Homogeneous competition TR-FRET assay for inhibitor screening.
  • Available in 100-test and 500-test kit sizes; larger quantities by inquiry.
  • Includes Europium-chelate labeled TL1A, FA-labeled DR3, control antibody, and TR-FRET buffers.
  • Assay time 0.5-1 hour; typical completion in 1 hour.
  • Store unopened kit at 2-8 °C; opened kit stable for 30 days.
  • Validated with antibody inhibitors; demonstrated IC50 values in the low nanomolar range.
Catalog No. NC3731147 Supplier ACROBiosystems Supplier No. FRT02100TESTS
Human TL1A-DR3 Inhibition Kit (TR-FRET) is a homogeneous, no-wash competition assay for rapid screening of inhibitors that block TL1A binding to DR3. The assay uses time-resolved fluorescence resonance energy transfer for sensitive, high-throughput detection and produces results within 0.5-1 hour.
  • Homogeneous competition TR-FRET assay for inhibitor screening.
  • Available in 100-test and 500-test kit sizes; larger quantities by inquiry.
  • Includes Europium-chelate labeled TL1A, FA-labeled DR3, control antibody, and TR-FRET buffers.
  • Assay time 0.5-1 hour; typical completion in 1 hour.
  • Store unopened kit at 2-8 °C; opened kit stable for 30 days.
  • Validated with antibody inhibitors; demonstrated IC50 values in the low nanomolar range.