Learn More
Sino Biological Recombinant Human TL1A/TNFSF15 Protein (His Tag)
Supplier:  Sino Biological 17049H07H1
Encompass_Preferred
A DNA sequence encoding the human TNFSF15 (NP_005109.2) (Leu72-Leu251) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-224-7178