Learn More
ACROBiosystems ACROBiosystems Human TL1A / TNFSF15 Protein, His Tag (MALS verified)
Catalog No. 502106356
Encompass_Preferred
Recombinant Protein;100UG;Human TL1A, His Tag (TLA-H5243) is expressed from human 293 cells (HEK293). It contains AA Leu 72- Leu 251 (Accession # O95150-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 22.4 kDa. The protein migrates as 26 kDa and 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DcR3, Fc Tag (Cat. No. TNB-H5255) at 5 μg/mL (100 μL/well) can bind Human TL1A, His Tag (Cat. No. TLA-H5243) with a linear range of 0.3-2 ng/mL (QC tested).
Catalog No. 50-210-6356 Supplier ACROBiosystems Supplier No. TLAH5243100UG
Recombinant Protein;100UG;Human TL1A, His Tag (TLA-H5243) is expressed from human 293 cells (HEK293). It contains AA Leu 72- Leu 251 (Accession # O95150-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 22.4 kDa. The protein migrates as 26 kDa and 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DcR3, Fc Tag (Cat. No. TNB-H5255) at 5 μg/mL (100 μL/well) can bind Human TL1A, His Tag (Cat. No. TLA-H5243) with a linear range of 0.3-2 ng/mL (QC tested).