Learn More
Raybiotech Inc Human TLR2 ELISA Kit
Supplier:  Raybiotech Inc ELHTLR21
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.32 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human TLR2.
Catalog No. 50-194-9788