Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TLR-4 Recombinant Protein, PeproTech®

Catalog No. 16006100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16006100UG 100 μg
160-06-10UG 10 μg
160-06-1MG 1 mg
16006250UG 250 μg
16006500UG 500 μg
160-06-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 16006100UG Supplier Gibco™ Supplier No. 16006100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TLR-4 is a 69.3 kDa glycoprotein containing 609 amino acid residues of the TLR-4 extracellular domain. As a result of glycosylation, Recombinant Human TLR-4 migrates with an apparent molecular mass of approximately 90-100 kDa by SDS-PAGE gel, under reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TLR4 is member of the Toll like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. Mammalian cells respond to LPS by activating TLR4. TLR4 belongs to the multi-protein complex of lipopolysaccharide (LPS) receptor, containing CD14, LY96 and TLR4, and is involved in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria. TLR4 aids in the recognition of pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. Mutations in the TLR4 gene have been associated with differences in LPS responsiveness. Also, several transcript variants of the TLR4 gene have been found, but the protein coding potential of most of them is uncertain. TLR4 is expressed by peripheral blood monocytes and a small population of B-cells and is also expressed in human placenta. Studies with TLR4-deficient mice indicate that the main ligand for TLR is lipopolysaccharide. Consequently, these mice also showed increased susceptibility to Gram-negative sepsis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00206
For Use With (Application) Control, ELISA Standard, Binding Assay, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 7099
Molecular Weight (g/mol) 69.3 kDa
Name Human TLR-4
Purification Method Purified
Quantity 100 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias ARMD10; CD284; EGK_07381; homolog of Drosophila toll; hToll; lipopolysaccharide response; Lps; Ly87; Ran/M1; Rasl2-8; Tlr4; TLR-4; TOLL; toll like receptor 4; toll/interleukin-l receptor (TIR) domain; toll4; toll-like receptor 4; toll-like receptor 4 immunity-related protein; Toll-like receptor 4-like protein; Toll-like receptor4 protein
Common Name TLR4 (CD284)
Gene Symbol TLR4
Biological Activity Determined by its ability to bind rhMD-2 in. a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence ESWEPCVEVV PNITYQCMEL NFYKIPDNLP FSTKNLDLSF NPLRHLGSYS FFSFPELQVL DLSRCEIQTI EDGAYQSLSH LSTLILTGNP IQSLALGAFS GLSSLQKLVA VETNLASLEN FPIGHLKTLK ELNVAHNLIQ SFKLPEYFSN LTNLEHLDLS SNKIQSIYCT DLRVLHQMPL LNLSLDLSLN PMNFIQPGAF KEIRLHKLTL RNNFDSLNVM KTCIQGLAGL EVHRLVLGEF RNEGNLEKFD KSALEGLCNL TIEEFRLAYL DYYLDDIIDL FNCLTNVSSF SLVSVTIERV KDFSYNFGWQ HLELVNCKFG QFPTLKLKSL KRLTFTSNKG GNAFSEVDLP SLEFLDLSRN GLSFKGCCSQ SDFGTTSLKY LDLSFNGVIT MSSNFLGLEQ LEHLDFQHSN LKQMSEFSVF LSLRNLIYLD ISHTHTRVAF NGIFNGLSSL EVLKMAGNSF QENFLPDIFT ELRNLTFLDL SQCQLEQLSP TAFNSLSSLQ VLNMSHNNFF SLDTFPYKCL NSLQVLDYSL NHIMTSKKQE LQHFPSSLAF LNLTQNDFAC TCEHQSFLQW IKDQRQLLVE VERMECATPS DKQGMPVLSL NITCQMNKT
Content And Storage -20°C
Activity Determined by its ability to bind rhMD-2 in a functional ELISA.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.