Learn More
Invivogen Human TLR3/7/8/9 Agonist kit
SDP
Catalog No. NC1077606
Encompass
This Toll-like Receptor (TLR) agonist kit represents a convenient and economical tool to study the stimulation of the TLRs. This kit contains several known TLR agonists and allows to perform up to 100 tests (in a 96-well plate).
Catalog No. NC1077606 Supplier Invivogen Supplier No. TLRLKIT3HW3
This Toll-like Receptor (TLR) agonist kit represents a convenient and economical tool to study the stimulation of the TLRs. This kit contains several known TLR agonists and allows to perform up to 100 tests (in a 96-well plate).