Learn More
Sino Biological Human TLR3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 106769T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TLR3 / CD283, and purified by antigen affinity chromatography.
Catalog No. 50-237-2751