Learn More
Sino Biological Human TLR3 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 10190H08B1
Encompass_Preferred
A DNA sequence encoding the Human TLR3 (AAH59372.1) (Lys27-Ser711) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1649