Learn More
Invivogen Human TLR9-expressing HEK293 cells, 3-7 x 10e6 cells
SDP
Supplier:  Invivogen HKBHTLR9
Encompass
HEK-Blue hTLR9 cells overexpress the human TLR9 (hTLR9) gene. They also express a SEAP (secreted embryonic alkaline phosphatase) reporter gene under the control of an NF-κB inducible promoter comprised of an IFN-β minimal promoter fused to five NF-κB and AP-1 binding sites. Levels of SEAP in the supernatant can be easily determined with HEK-Blue™ Detection, a cell culture medium that allows real-time detection of SEAP activity.
Catalog No. NC0027929