Learn More
Invivogen INVIVOGEN THPD-HTLR9, Specialized Reporter Cell Line, Overexpresses Human TLR9, Measures NF-κB and IRF Activation, 3-7 x 10^6 cells
SDP
Supplier:  Invivogen THPDHTLR9
Encompass
The INVIVOGEN THPD-HTLR9 is a specialized THP1-Dual™ hTLR9 cell line that facilitates the investigation of human TLR9 signaling pathways. This product allows researchers to study innate immune responses using cells that stably overexpress human TLR9, exhibiting strong responsiveness to various TLR9 agonists. Its dual-reporter system for IRF and NF-κB pathway monitoring sets it apart from other models, making it ideal for sensitive and reliable assessment of TLR9 activation effects.
Catalog No. NC2006554