Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TLX1 Synthetic Peptide

Catalog No. PEP1596 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1596 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1596 Supplier Invitrogen™ Supplier No. PEP1596
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 16 amino acid peptide near the amino terminus of human TLX1. PEP-1596 can be used as a blocking peptide with polyclonal antibody PA5-34553. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37°C.

The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX1, also called homeobox11 (HOX11), as it is located outside of the four mammalian Hox clusters, is a DNA-binding nuclear transcription factor. TLX1 encodes a homeobox-domain containing protein and is containing a glycine and proline-rich amino terminus. TLX1 is required for maintenance of the developing spleen and cell survival. TLX1 is highly expressed in T-cell leukemia and lymphoid neoplasias as a result of translocation. TLX1-deficient mice have no spleen.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 3195
Species Human
Purification Method purified
Accession Number P31314
Common Name TLX1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias homeo box-11 (T-cell leukemia-3 associated breakpoint, homologous to Drosophila Notch); homeobox protein Hox11; homeobox protein Hox-11; homeobox TLX-1; HOX11; Hox-11; LOW QUALITY PROTEIN: T-cell leukemia homeobox protein 1; MGC163402; proto-oncogene TCL-3; RGD1563655; SI:dZ46P4.1; T cell leukemia homeobox 1; T cell leukemia, homeobox 1; T-cell leukemia homeobox 1; T-cell leukemia homeobox protein 1; T-cell leukemia, homeobox 1; T-cell leukemia/lymphoma protein 3; TCL3; TLX1; Tlx-1; transcription factor; ztlx-1
Gene Symbol TLX1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.