Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TM4SF18 (aa 68-91) Control Fragment Recombinant Protein

Catalog No. RP109789
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109789 Supplier Invitrogen™ Supplier No. RP109789
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (38%), Rat (38%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144874. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Transmembrane 4 L Six Family Member 18 (TM4SF18) is a gene located on chromosome 11q12.3 that belongs to the transmembrane 4 L six superfamily, a group of proteins characterized by the presence of four transmembrane domains. TM4SF18 is expressed in various tissues and is involved in cellular processes such as cell adhesion, migration, and proliferation. As part of the tetraspanin family, TM4SF18 proteins play a role in organizing cellular membranes and facilitating interactions between cell surface receptors, impacting signaling pathways crucial for maintaining cellular functions and tissue homeostasis. Although TM4SF18's specific biological roles are still being elucidated, it has been implicated in tumorigenesis, where altered expression may influence cancer progression by modulating cell-cell communication and migration. Research into TM4SF18 aims to uncover its precise functions in health and disease, particularly its potential involvement in cancer biology and as a target for therapeutic interventions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96CE8
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 116441
Name Human TM4SF18 (aa 68-91) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AAB5; L6D; TM4SF18; transmembrane 4 L six family member 18; transmembrane 4 L6 family member 18
Common Name TM4SF18
Gene Symbol TM4SF18
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LVLENNNNYKCCQSENCSKKYVTL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.