Learn More
BIOHIPPO Human TMAO Microsample
SDP
Catalog No. 5000314100
Encompass
Human TMAO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMAO in serum plasma cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 mol/L with a sensitivity of 0 065 mol/L The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TMAO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TMAO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TMAO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14100 Supplier BIOHIPPO Supplier No. BHE15210709
Human TMAO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMAO in serum plasma cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 mol/L with a sensitivity of 0 065 mol/L The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TMAO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TMAO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TMAO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color