Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TMC6 Synthetic Peptide

Catalog No. PEP0624 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0624 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0624 Supplier Invitrogen™ Supplier No. PEP0624
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 14 amino acids near the amino terminus of human EVER1. PEP-0624 can be used as a blocking peptide with polyclonal antibody PA5-20504.

TMC6 interacts with, and forms a complex with the zinc transporter 1, SLC30A1, and localizes to the endoplasmic reticulum, nuclear membrane and Golgi apparatus. The inactivation of the TMC6 gene has been implicated in the skin disorder epidermodysplasia verruciformis (EV), an autosomal recessive dermatosis characterized by abnormal susceptibility to human papillomaviruses (HPVs) and a high rate of progression to squamous cell carcinoma on sun-exposed skin. EV is caused by mutations in either of two adjacent genes located on chromosome 17q25.3. TMC6 encodes integral membrane proteins that localize to the endoplasmic reticulum and are predicted to form transmembrane channels. This gene encodes a transmembrane channel-like protein with 10 transmembrane domains and 2 leucine zipper motifs. Diseases associated with TMC6 include Epidermodysplasia Verruciformis and Superficial Mycosis.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 11322
Species Human
Purification Method purified
Accession Number Q7Z403
Common Name TMC6
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias D11Ertd204e; epidermodysplasia verruciformis 1; epidermodysplasia verruciformis protein 1; EV1; EVER1; EVIN1; expressed in activated T/LAK lymphocytes; LAK-4P; protein LAK-4; Tmc6; transmembrane channel like 6; transmembrane channel-like gene family 6; transmembrane channel-like protein 6
Gene Symbol TMC6
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.