Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TMEM123 (aa 36-93) Control Fragment Recombinant Protein

Catalog No. RP108478
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108478 Supplier Invitrogen™ Supplier No. RP108478
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (34%), Rat (34%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111556. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Transmembrane protein 123, or Tmem123, is a single-pass type I membrane protein and belongs to the CD164 family. Tmem123 acts as a novel marker for DC maturation. DCs are also known as professional antigen-presenting cells. It is reported that the Tmem123 molecule may be closely associated with the cell surface expression of CD40. It was also reported previously that Tmem123 protein may also be responsible for oncotic cell death, which is characterized by cell swelling, organelle swelling, vacuolization, increased membrane permeability and cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N131
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 114908
Name Human TMEM123 (aa 36-93) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2310075C12Rik; KCT3; KCT-3; keratinocytes associated transmembrane protein 3; keratinocytes-associated transmembrane protein 3; Porimin; PORMIN; pro oncosis receptor inducing membrane injury; pro-oncosis receptor inducing membrane injury; PSEC0111; RGD1305625; serine/threonine-rich receptor; TMEM123; Transmembrane protein 123; UNQ641/PRO1271
Common Name TMEM123
Gene Symbol TMEM123
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ANIENSGLPHNSSANSTETLQHVPSDHTNETSNSTVKPPTSVASDSSNTTVTTMKPTA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.