Learn More
BIOHIPPO Human TMEM176B Microsample
SDP
Catalog No. 5000314377
Encompass
Human TMEM176B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMEM176B in tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 62 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TMEM176B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TMEM176B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TMEM176B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14377 Supplier BIOHIPPO Supplier No. BHE15210813
Human TMEM176B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMEM176B in tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 62 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TMEM176B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TMEM176B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TMEM176B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color