Learn More
Abcam Recombinant Human Tmem27 protein, 2UG
Supplier:  Abcam AB1327082UG
Encompass_Preferred
Recombinant Human Tmem27 protein is a Human Full Length protein, in the 1 to 222 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: TMEM27, UNQ679/PRO1312, CLTRN, Collectrin, Transmembrane protein 27
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2323173