Learn More
BIOHIPPO Human TMEM2L Microsample
SDP
Catalog No. 5000313813
Encompass
Human TMEM2L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMEM2L in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TMEM2L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TMEM2L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TMEM2L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13813 Supplier BIOHIPPO Supplier No. BHE15211121
Human TMEM2L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMEM2L in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TMEM2L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TMEM2L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TMEM2L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color