Learn More
BIOHIPPO Human TMEM48 Microsample
SDP
Catalog No. 5000315738
Encompass
Human TMEM48 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMEM48 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TMEM48 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TMEM48 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TMEM48 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15738 Supplier BIOHIPPO Supplier No. BHE15210639
Human TMEM48 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TMEM48 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TMEM48 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TMEM48 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TMEM48 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color