Learn More
Sino Biological Human TMIGD1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 51003M49HB100
Encompass_Preferred
A DNA sequence encoding the human TMIGD1 (NP_996663.1) (Met1-Pro220) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-237-4518