Learn More
Sino Biological Anti-CD28H/IGPR-1/TMIGD2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13820T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TMIGD2 (rh TMIGD2; Catalog#13820-H08H; NP_653216.2; Met1-Gly150). TMIGD2 specific IgG was purified by Human TMIGD2 affinity chromatography.
Catalog No. 50-255-4180