Learn More
Sino Biological Recombinant Human CD28H/IGPR-1/TMIGD2 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 13820H27HB100
Encompass_Preferred
A DNA sequence encoding the human TMIGD2 (NP_001162597.1) (Met1-Gly150) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4182