Learn More
Abcam Human TNF alpha ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1814211X96TESTS
Encompass_Preferred
Human TNF alpha ELISA kit is a single-wash 90-min SimpleStep ELISA used to quantify Human TNFα with a sensitivity of 4.32 pg/mL. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric assay - 450nm readout; works on any plate reader - Different formats for different needs: 10x 96-well plates for bulk orders and 384-well for higher throughput - Broad sample compatibility – works with CSF, serum, plasma, cell culture supernatants, and other biological fluids - Cited in over 250 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1381431