Learn More
ACROBiosystems Biotinylated Human TNF-alpha Protein, His,Avitag™, active trimer (MALS verified)
Catalog No. 502331065
Encompass_Preferred
Biotinylated Human TNF-alpha, His,Avitag (TNA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # NP_000585.2).
Catalog No. 50-233-1065 Supplier ACROBiosystems Supplier No. TNAH82E3200UG
Biotinylated Human TNF-alpha, His,Avitag (TNA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # NP_000585.2).