Learn More
Invitrogen™ Human TNF-beta Recombinant Protein
Description
RP-75741 contains purified recombinant human TNF beta protein with an N-terminal hexahistidine tag. RP-75741 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P01374 |
| For Use With (Application) | Control, Western Blot |
| Formulation | 10 mM Bis-tris propane with 50% glycerol, 62.5 mM NaCl and no preservative; pH 9 |
| Gene ID (Entrez) | 4049 |
| Name | Human TNF-beta |
| Quantity | 1 mg |
| Source | E. Coli |
| Regulatory Status | RUO |
| Gene Alias | DAMA-25N12.13; hlb382; hypothetical protein; LT; LT[a]; LT-[a]; LTA; LTalpha; LT-alpha; Ltx; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); lymphotoxin-alpha; lymphotoxin-alpha; lymphotoxin alpha; TNF b; TNF beta; TNF superfamily, member 1; TNF β; Tnfb; TNFbeta; TNF-beta; TNF-N; Tnfsf1; Tnfsf1b; TNFβ; TNLG1E; tumor necrosis factor beta; tumor necrosis factor ligand 1 E; tumor necrosis factor ligand superfamily member 1 |
| Common Name | TNF beta |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.