Learn More
Sino Biological Human TNF ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological 102334T32100
Encompass_Preferred
Each vial contains 78 ng of recombinant Human TNF-alpha / TNFA / TNFSF2. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 1500 pg/mL is recommended.
Catalog No. 50-237-4025