Learn More
Sino Biological Anti-TNF-alpha Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10602MM01F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNF-alpha (rh TNF-alpha; Catalog#10602-HNAE; NP_000585.2; Val 77-Leu 233) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1259