Learn More
Sino Biological Anti-TNF-alpha Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10602RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TNF-alpha / TNFA (rh TNF-alpha / TNFA; Catalog#10602-HNAE; NP_000585.2; Val77-Leu233). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8796