Learn More
Sino Biological Recombinant Human TNF-alpha Protein, GMP grade, HPLC-verified
Supplier:  Sino Biological GMP10602HNAE1
Encompass_Preferred
A DNA sequence encoding the human TNF-α soluble form (NP_000585.2) (Val77-Leu233) was expressed with an initial Met at the N-terminus.
Catalog No. 50-237-1733