Learn More
Biomatik Corporation Human TNF Receptor Associated Factor 6 (TRAF6) ELISA Kit
Supplier:  Biomatik Corporation EKU07741
Encompass_Preferred
Human TNF Receptor Associated Factor 6 (TRAF6) ELISA Kit for detecting TRAF6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y4K3. Gene ID: 7189. Target Synonyms: RNF85; E3 ubiquitin-protein ligase TRAF6; Interleukin-1 signal transducer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8728