Learn More
Meso Scale Discovery HUMAN TNF-RI ASSAY, 5 POINT, Electrochemiluminescence (ECL) Immunoassay, 96-well plate, Dynamic range 0.15 - 1,000 pg/mL, LLOD 0.15 pg/mL
SDP
Supplier:  Meso Scale Discovery K151AHQK2
Encompass
HUMAN TNF-RI ASSAY, 5 POINT, Electrochemiluminescence (ECL) Immunoassay, 96-well plate, Dynamic range 0.15 - 1,000 pg/mL, LLOD 0.15 pg/mL
Catalog No. NC3343253