Learn More
Sino Biological Human TNFAIP8 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 14559MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNFAIP8 (rh TNFAIP8; Catalog#14559-H07E; O95379; Met1-Ile198). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1403