Learn More
ACROBiosystems Human TNFR2 / CD120b / TNFRSF1B Protein, Fc Tag (MALS verified)
Catalog No. 502330918
Encompass_Preferred
Human TNFR2 / CD120b / TNFRSF1B Protein, Fc Tag (TN2-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asp 257 (Accession # P20333-1).
Catalog No. 50-233-0918 Supplier ACROBiosystems Supplier No. TN2H525350UG
Human TNFR2 / CD120b / TNFRSF1B Protein, Fc Tag (TN2-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asp 257 (Accession # P20333-1).