Learn More
Sino Biological Recombinant Human TNFR-2 / CD120b Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10417H27HB20
Encompass_Preferred
A DNA sequence encoding the human TNFRSF1B (NP_001057.1) (Met1-Asp257) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-228-2310