Learn More
ACROBiosystems ACROBiosystems Human TNFR2 / CD120b / TNFRSF1B Protein, His Tag
Catalog No. 502106758
Encompass_Preferred
Recombinant Protein;1MG;Human TNFR2, His Tag (TN2-H5227) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asp 257 (Accession # AAA36755).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.0 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human TNF-alpha, Tag Free, low endotoxin (Cat. No. TNA-H4211) at 5 μg/mL (100 μL/well) can bind Human TNFR2, His Tag (Cat. No. TN2-H5227) with a linear range of 5-39 ng/mL (QC tested).;Immobilized ActiveMax Human TNF-beta, Tag Free (Cat. No. TNB-H5214) at 5 μg/mL (100 μL/well) can bind Human TNFR2, His Tag (Cat. No. TN2-H5227) with a linear range of 2-62 ng/mL (Routinely tested).
Catalog No. 50-210-6758 Supplier ACROBiosystems Supplier No. TN2H52271MG
Recombinant Protein;1MG;Human TNFR2, His Tag (TN2-H5227) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asp 257 (Accession # AAA36755).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.0 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human TNF-alpha, Tag Free, low endotoxin (Cat. No. TNA-H4211) at 5 μg/mL (100 μL/well) can bind Human TNFR2, His Tag (Cat. No. TN2-H5227) with a linear range of 5-39 ng/mL (QC tested).;Immobilized ActiveMax Human TNF-beta, Tag Free (Cat. No. TNB-H5214) at 5 μg/mL (100 μL/well) can bind Human TNFR2, His Tag (Cat. No. TN2-H5227) with a linear range of 2-62 ng/mL (Routinely tested).