Learn More
Sino Biological Anti-DR5/TRAIL R2 Antibody (HRP), Rabbit Polyclonal
Supplier:  Sino Biological 10465T16H100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TRAIL R2/CD262/TNFRSF10B (rh TRAIL R2/CD262/TNFRSF10B; Catalog#10465-H08H; NP_003833.4; Met1-Glu182). TRAIL R2/CD262/TNFRSF10B specific IgG was purified by Human TRAIL R2/CD262/TNFRSF10B affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1040