Learn More
Sino Biological Recombinant Human DR5/TRAIL R2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10465H08H1
Encompass_Preferred
A DNA sequence encoding the human TNFRSF10B (NP_003833.4) extracellular domain (Met1-Glu182) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5288