Learn More
Sino Biological Recombinant Human DcR2/TRAIL R4 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 10413H02H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF10D (NP_003831.2) (Met1-His211) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5311