Learn More
Sino Biological Recombinant Human RANK/TNFRSF11A Protein (hFc Tag), HPLC-verified 1
Supplier:  Sino Biological 16078H02H1
Encompass_Preferred
A DNA sequence encoding the human TNFRSF11A (NP_001257879.1) (Met1-Pro212) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6768