Learn More
Sino Biological Anti-BAFFR/TNFRSF13C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200139T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human BAFF R / TNFRSF13C fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3974