Learn More
Sino Biological Recombinant Human BCMA Protein (Site-Specific AF 488 Conjugation, ECD, His Tag)
Catalog No. 502992927
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Ala54) of human TNFRSF17 (NP_001183.2) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with AF 488 (Excitation Max.= 495 nm, Emission Max.= 519 nm).
Catalog No. 50-299-2927 Supplier Sino Biological Supplier No. 10620H86HSD100
A DNA sequence encoding the extracellular domain (Met1-Ala54) of human TNFRSF17 (NP_001183.2) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with AF 488 (Excitation Max.= 495 nm, Emission Max.= 519 nm).