Learn More
Sino Biological Anti-BCMA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10620RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BCMA / TNFRSF17 / CD269 (rh BCMA / TNFRSF17 / CD269 ; Catalog#10620-H15H; NP_001183.2; Met1-Ala54). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1312