Learn More
Sino Biological Recombinant Human BCMA Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10620H49HB100
Encompass_Preferred
A DNA sequence encoding the Human TNFRSF17 (NP_001183.2) (Met1-Ala54) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5238