Learn More
Sino Biological Recombinant Human BCMA Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10620H02H50
Encompass_Preferred
A DNA sequence encoding the human TNFRSF17 (NP_001183.2) (Met1-Ala54) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5242