Learn More
Sino Biological Anti-GITR/TNFRSF18/AITR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13643RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GITR / TNFRSF18 (rh GITR / TNFRSF18; Catalog#13643-H08H; NP_683700.1; Met1-Glu161). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4199