Learn More
Sino Biological Human TNFRSF18 Protein 20ug (Sino Biological)
Catalog No. 502374195
Encompass_Preferred
A DNA sequence encoding the Human TNFRSF18 (NP_683700.1) (Met1-Glu161) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-4195 Supplier Sino Biological Supplier No. SEKA109475
A DNA sequence encoding the Human TNFRSF18 (NP_683700.1) (Met1-Glu161) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.