Learn More
Sino Biological Human TNFRSF19 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 204104T44100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNFRSF19/TROY (rh TNFRSF19/TROY; Catalog#13387-H08H; Q9NS68-2; Met1-Leu170). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4299