Learn More
Sino Biological Anti-TROY/TNFRSF19 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106043T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TROY / TNFRSF19, and purified by antigen affinity chromatography.
Catalog No. 50-240-8663