Learn More
Sino Biological Anti-TNFR1/CD120a Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10872MM09TH100
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells. Then conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-0476